Learn More
Medchemexpress LLC Leukotriene D4 | 73836-78-9 | 98.5% | 496.66 | 10 UG
SDP
Catalog No. 5000436594
Encompass_Preferred
Leukotriene D4 is a potent bronchoconstrictor with potential for asthma research. It induces edema and increases capillary permeability, and is for research use only.
  • Potent bronchoconstrictor
  • Potential for asthma research
  • Induces edema
  • Increases capillary permeability
Catalog No. 50-004-36594 Supplier Medchemexpress LLC Supplier No. HY11345610UG
Leukotriene D4 is a potent bronchoconstrictor with potential for asthma research. It induces edema and increases capillary permeability, and is for research use only.
  • Potent bronchoconstrictor
  • Potential for asthma research
  • Induces edema
  • Increases capillary permeability