Learn More
Medchemexpress LLC Leukotriene D4 | 73836-78-9 | 98.5% | 496.66 | 50 UG
SDP
Catalog No. 5000437799
Encompass_Preferred
Leukotriene D4 is a potent bronchoconstrictor with potential for asthma research. It induces edema and increases capillary permeability.
  • Potent bronchoconstrictor for research applications.
  • Potential for asthma research.
  • Induces edema and increases capillary permeability.
  • Demonstrates significant edematous response and dye extravasation in animal models.
Catalog No. 50-004-37799 Supplier Medchemexpress LLC Supplier No. HY11345650UG
Leukotriene D4 is a potent bronchoconstrictor with potential for asthma research. It induces edema and increases capillary permeability.
  • Potent bronchoconstrictor for research applications.
  • Potential for asthma research.
  • Induces edema and increases capillary permeability.
  • Demonstrates significant edematous response and dye extravasation in animal models.