Learn More
Cayman Chemical Leukotriene D4d5
SDP
Catalog No. 502889439
Encompass_Preferred
An internal standard for the quantification of LTD4 by GC- or LC-MS
Catalog No. 50-288-9439 Supplier Cayman Chemical Supplier No. 1000619950 UG
An internal standard for the quantification of LTD4 by GC- or LC-MS