Learn More
Enzo Life Sciences Leukotriene E4-20,20,20-d3 (50µg)
SDP
Supplier:  Enzo Life Sciences BMLLE5040050
Encompass_Preferred
Alternative name: LTE4-d3. Formulation: Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1148