Learn More
Enzo Life Sciences Leukotriene E4 (50µg). CAS: 75715-89-8
SDP
Supplier:  Enzo Life Sciences BMLLE0040050
Encompass_Preferred
Less potent than LTD4 in smooth muscle contractile activity and mucus secretion in airways. Reduces myocardial contractility and coronary blood flow. Induces long lasting hypotension and airway hyperresponsiveness. Partial agonist at LTD4 receptors. LTE4 solely activates a CysLT1 receptor. Biosynthesis from LTD4 by a dipeptidase. Alternative name: LTE4. Formulation: Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1146