Learn More
Cayman Chemical Leukotriene E4d5
SDP
Catalog No. 502887866
Encompass_Preferred
An internal standard for the quantification of LTE4 by GC- or LC-MS
Catalog No. 50-288-7866 Supplier Cayman Chemical Supplier No. 1000785850 UG
An internal standard for the quantification of LTE4 by GC- or LC-MS