Learn More
Bachem Leupeptin
SDP
Supplier:  Bachem 4013467.0025
Encompass_Preferred
Leupeptin, Ac-Leu-Leu-Arg-aldehyde · sulfate salt, 103476-89-7, 2 C20H38N6O4 · H2SO4, Mr 951.20, 25mg. A reversible compeptitive protease inhibitor, which was shown to inhibit cathepsins B, H, L, and S, calpain and trypsin. Ac-LLR-CHO inhibits the trypsin-like activity of the proteasome endopeptidase complex. CAS Number (hemisulfate): 103476-89-7.
Catalog No. 50-260-506