Learn More
MilliporeSigma Leupeptin 100mg
Catalog No. 5056849
Encompass
Leupeptin 100mg. Storage: Maintain dry at -20ºC for up to 18 months after date of receipt. Store reconstituted product in aliquots at -20ºC for up to 6 months. Avoid repeated thaw freeze cycles.
Catalog No. 50-568-49 Supplier MilliporeSigma Supplier No. EI8
Leupeptin 100mg. Storage: Maintain dry at -20ºC for up to 18 months after date of receipt. Store reconstituted product in aliquots at -20ºC for up to 6 months. Avoid repeated thaw freeze cycles.