Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leupeptin, >96.5%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15155301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15155301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15155301 Supplier MP Biomedicals, Inc Supplier No. 0215155301

missing translation for 'validMainframeMsg'

Specifications

Purity >96.5%
Assay >96.5%
CAS 103476-89-7
Color White
Format Powder
Molecular Weight (g/mol) 427.57
Product Type Leupeptin Hemisulfate
Quantity 1 mg
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.