Learn More
Research Products International Corp Leupeptin Hemisulfate, 1 Milligram
SDP
Catalog No. 501368324
Encompass_Preferred
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM
Catalog No. 50-136-8324 Supplier Research Products International Corp Supplier No. L220350.001
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM