Learn More
Alkali Scientific Leupeptin Hemisulfate, 100 Milligrams
SDP
Catalog No. 5000659935
Encompass_Preferred
Leupeptin Hemisulfate is a protease inhibitor commonly used in biochemical research to block the activity of a broad range of proteases. This reagent, supplied in a 100 mg quantity, is essential for maintaining the integrity of protein samples by preventing proteolytic degradation during experimental procedures. Leupeptin inhibits both serine and cysteine proteases, making it particularly useful in studies involving cell culture, enzyme activity, and protein analysis. This chemical is often utilized in research involving enzyme inhibition, purification procedures, and protein quantification in areas like molecular biology, cancer research, and immunology. Leupeptin is typically used at concentrations of 10 to 50 µM for effective protease inhibition.
Catalog No. 50-006-59935 Supplier Alkali Scientific Supplier No. LEU004CP
Leupeptin Hemisulfate is a protease inhibitor commonly used in biochemical research to block the activity of a broad range of proteases. This reagent, supplied in a 100 mg quantity, is essential for maintaining the integrity of protein samples by preventing proteolytic degradation during experimental procedures. Leupeptin inhibits both serine and cysteine proteases, making it particularly useful in studies involving cell culture, enzyme activity, and protein analysis. This chemical is often utilized in research involving enzyme inhibition, purification procedures, and protein quantification in areas like molecular biology, cancer research, and immunology. Leupeptin is typically used at concentrations of 10 to 50 µM for effective protease inhibition.