Learn More
Ubpbio A reversible inhibitor of serine and cysteine proteases. CAS# 103476-89-7, > 98% purity
Catalog No. 502339322
Encompass_Preferred
Leupeptin hemisulfate (Ac-Leu-Leu-argininal) is a reversible inhibitor of serine and cysteine proteases, including plasmin, trypsin, papain, kallikrein, calpain and cathepsin B. A widely used protease inhibitor and typical working concentration is 10 - 100 μM. 
Catalog No. 50-233-9322 Supplier Ubpbio Supplier No. P105710
Add to Cart
Add to Cart
Leupeptin hemisulfate (Ac-Leu-Leu-argininal) is a reversible inhibitor of serine and cysteine proteases, including plasmin, trypsin, papain, kallikrein, calpain and cathepsin B. A widely used protease inhibitor and typical working concentration is 10 - 100 μM. 