Learn More
Research Products International Corp RPI Leupeptin Hemisulfate, 50 Milligrams
SDP
Catalog No. 501977374
Encompass_Preferred
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM
Catalog No. 50-197-7374 Supplier Research Products International Corp Supplier No. L220350.050
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM