Learn More
Research Products International Corp Leupeptin Hemisulfate, 250 Milligrams
SDP
Catalog No. 501368326
Encompass_Preferred
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM
Catalog No. 50-136-8326 Supplier Research Products International Corp Supplier No. L220350.25
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM