Learn More
Alkali Scientific Leupeptin Hemisulfate, 250 Milligrams
SDP
Catalog No. 5000659907
Encompass_Preferred
Leupeptin Hemisulfate is a potent protease inhibitor widely used in biochemical and molecular biology research to prevent protein degradation. It specifically inhibits serine and cysteine proteases, protecting proteins from enzymatic breakdown during extraction and purification processes. This reagent is commonly included in lysis buffers and protein extraction protocols to maintain protein integrity and preserve enzymatic activity. Leupeptin is particularly useful in studies involving proteomics, enzyme characterization, and cell signaling pathways. Its stability and effectiveness make it a standard component in laboratory workflows where accurate protein analysis and preservation are essential.
Catalog No. 50-006-59907 Supplier Alkali Scientific Supplier No. LEU006CP
Leupeptin Hemisulfate is a potent protease inhibitor widely used in biochemical and molecular biology research to prevent protein degradation. It specifically inhibits serine and cysteine proteases, protecting proteins from enzymatic breakdown during extraction and purification processes. This reagent is commonly included in lysis buffers and protein extraction protocols to maintain protein integrity and preserve enzymatic activity. Leupeptin is particularly useful in studies involving proteomics, enzyme characterization, and cell signaling pathways. Its stability and effectiveness make it a standard component in laboratory workflows where accurate protein analysis and preservation are essential.