Learn More
TARGETMOL CHEMICALS INC Leupeptin Hemisulfate 25MG
SDP
Catalog No. 502876354
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Leupeptin hemisulfate is a protease inhibitor with cell membrane-permeable, reversible, competitive, and oral activities. Leupeptin hemisulfate inhibits the activity of Cathepsin B, Cathepsin H, and Cathepsin L, and blocks fusion of amphipathic lysosomes. Leupeptin hemisulfate also has anti-inflammatory activity. Purity ≥98%
Catalog No. 50-287-6354 Supplier TARGETMOL CHEMICALS INC Supplier No. T656425MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Leupeptin hemisulfate is a protease inhibitor with cell membrane-permeable, reversible, competitive, and oral activities. Leupeptin hemisulfate inhibits the activity of Cathepsin B, Cathepsin H, and Cathepsin L, and blocks fusion of amphipathic lysosomes. Leupeptin hemisulfate also has anti-inflammatory activity. Purity ≥98%