Learn More
Gold Biotechnology Inc Leupeptin Hemisulfate 100 mg
SDP
Catalog No. NC2943371
Encompass_Preferred
Leupeptin hemisulfate (Ac-Leu-Leu-argininal) is a reversible inhibitor of serine and cysteine endopeptidases. Leupeptin hemisulfate is capable of inhibiting plasmin, trypsin, papain, kallikrein, calpain and cathepsin B. No inhibition has been found with pepsin, cathepsins A and D, elastase, renin or chymotrypsin.
Catalog No. NC2943371 Supplier Gold Biotechnology Inc Supplier No. L010100
Leupeptin hemisulfate (Ac-Leu-Leu-argininal) is a reversible inhibitor of serine and cysteine endopeptidases. Leupeptin hemisulfate is capable of inhibiting plasmin, trypsin, papain, kallikrein, calpain and cathepsin B. No inhibition has been found with pepsin, cathepsins A and D, elastase, renin or chymotrypsin.