Learn More
Selleck Chemical LLC Leupeptin Hemisulfate
SDP
Catalog No. 5000004396
Encompass_Preferred
Leupeptin Hemisulfate is a reversible inhibitor of serine and cysteine proteases It inhibits cathepsin B (Ki 6 nM) calpain (Ki 10 nM) trypsin (Ki 35 nM) plasmin (Ki 3 4 M) and kallikrein (Ki 19 M) and has no effect against chymotrypsin elastase renin or pepsin
Catalog No. 50-000-04396 Supplier Selleck Chemical LLC Supplier No. S738010MM/1ML
Leupeptin Hemisulfate is a reversible inhibitor of serine and cysteine proteases It inhibits cathepsin B (Ki 6 nM) calpain (Ki 10 nM) trypsin (Ki 35 nM) plasmin (Ki 3 4 M) and kallikrein (Ki 19 M) and has no effect against chymotrypsin elastase renin or pepsin