Learn More
Research Products International Corp Leupeptin Hemisulfate, 5 Milligrams
SDP
Catalog No. 50550413
Encompass_Preferred
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM
Catalog No. 50-550-413 Supplier Research Products International Corp Supplier No. L220350.005
Reversible inhibitor of cysteine and serine proteases resembling trypsin, Inhibits cathepsin B, kallikrein, papain, plasmin, thrombin and trypsin.
Working concentration: 1-10uM