Learn More
Abcam Leupeptin hemisulfate, serine/cysteine protease inhibitor, 25MG
Catalog No. 502706645
Encompass_Preferred
MW 426.6 Da, Purity >98%. Leupeptin hemisulfate, serine/cysteine protease inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6645 Supplier Abcam Supplier No. AB14140425MG
MW 426.6 Da, Purity >98%. Leupeptin hemisulfate, serine/cysteine protease inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement