Learn More
Indofine Chemical Leupeptin Hemisulp, 90%, 25 Mg
SDP
Catalog No. 50913527
Encompass_Preferred
LEUPEPTIN HEMISULPHATE 25 MG, Products for Life Science, Ac-Leu-Leu-argininal hemisulphate, 2 C20H38N6O4 . H2O4S, MW: 951.19, >90%, 103476-89-7, MFCD00037012
Catalog No. 50-913-527 Supplier Indofine Chemical Supplier No. BIO235 25 MG
LEUPEPTIN HEMISULPHATE 25 MG, Products for Life Science, Ac-Leu-Leu-argininal hemisulphate, 2 C20H38N6O4 . H2O4S, MW: 951.19, >90%, 103476-89-7, MFCD00037012