Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leupeptin, MP Biomedicals™

Catalog No. MP215155380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5mg
25mg
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215155380 100mg
MP215155305 5mg
MP215155325 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215155380 Supplier MP Biomedicals, Inc Supplier No. 0215155380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits trypsin-like proteases and some cysteine proteases. MP Biomedicals' Leupeptin is a modified tripeptide reversible protease inhibitor. Little to no inhibition is seen against pepsin, cathepsins A and D, and alpha-chymotrypsin.

When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.

Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.

TRUSTED_SUSTAINABILITY

Specifications

CAS 103476-89-7
Content And Storage Store at −20°C, desiccate, protect from light.
Format White Powder
Molecular Weight (g/mol) 427.57
Quantity 100mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.