Learn More
Sigma Aldrich Fine Chemicals Biosciences Leupeptin Ready Made Solut
Catalog No. 502900481
Encompass
Leupeptin is a tripeptide protease inhibitor that is naturally produced in Actinomyces / Streptomyces. Known also as N-acetyl-L-leucyl-L-leucyl-L-arginal leupeptin is a reversible competitive inhibitor of cysteine serine and thiol proteases.
Catalog No. 50-290-0481 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE01531ML
Leupeptin is a tripeptide protease inhibitor that is naturally produced in Actinomyces / Streptomyces. Known also as N-acetyl-L-leucyl-L-leucyl-L-arginal leupeptin is a reversible competitive inhibitor of cysteine serine and thiol proteases.