Learn More
Enzo Life Sciences Leupeptin (synthetic) (100mg). CAS: 103476-89-7
SDP
Catalog No. 502008220
Encompass_Preferred
Competitive and reversible inhibitor of serine and cysteine proteinases like calpain, porcine kallikrein, trypsin, plasmin, papain and cathepsin B. Does not inhibit chymotrypsin and thrombin. Also known to inhibit activation-induced programmed cell death and restoration of defective immune responses of HIV+ donors. Alternative name: Acetyl-L-leucyl-L-leucyl-L-argininal . hemisulfate. Formula: C20H38N6O4 . 0.5 H2SO4. MW: 426.6 . 49.0. Purity: ≥97% (HPLC). Appearance: White to off-white powder. MI: 14: 5456. Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -20°C. Handling: Protect from light and moisture.
Catalog No. 50-200-8220 Supplier Enzo Life Sciences Supplier No. ALX260009M100
Competitive and reversible inhibitor of serine and cysteine proteinases like calpain, porcine kallikrein, trypsin, plasmin, papain and cathepsin B. Does not inhibit chymotrypsin and thrombin. Also known to inhibit activation-induced programmed cell death and restoration of defective immune responses of HIV+ donors. Alternative name: Acetyl-L-leucyl-L-leucyl-L-argininal . hemisulfate. Formula: C20H38N6O4 . 0.5 H2SO4. MW: 426.6 . 49.0. Purity: ≥97% (HPLC). Appearance: White to off-white powder. MI: 14: 5456. Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -20°C. Handling: Protect from light and moisture.