Learn More
Selleck Chemical LLC Leuprolide Acetate 5mg 74381-53-6 Leuprorelin Acetate
SDP
Supplier:  Selleck Chemical LLC S37185MG
Encompass_Preferred
Leuprolide acetate (Leuprorelin) is a synthetic nonapeptide that is a potent gonadotropin-releasing hormone receptor (GnRHR) agonist used for diverse clinical applications, including the treatment of prostate cancer, endometriosis, uterine fibroids, central precocious puberty and in vitro fertilization techniques.
Catalog No. 50-194-8338