Learn More
Cayman Chemical LeuprorelInacetate 10mg
SDP
Catalog No. 502987827
Encompass_Preferred
A synthetic GnRH receptor agonist; long-acting; decreases circulating levels of gonadotropins and sex hormones
Catalog No. 50-298-7827 Supplier Cayman Chemical Supplier No. 2227510 MG
A synthetic GnRH receptor agonist; long-acting; decreases circulating levels of gonadotropins and sex hormones