Learn More
Andwin Scientific LEUR LOCK SYRINGE 10ML 100/PK
SDP
Catalog No. NC9001097
Encompass_Preferred
The Luer-Lock syringe is free of latex and does not use rubber or silicone lubricant. This product is ideal for IVF, embryo transfer, AI, chromatography and other several applications. The syringe has a volume capacity of 10 mL. It is sold in a pack of 100.
This item is not returnable. View return policy
Catalog No. NC9001097 Supplier Andwin Scientific Supplier No. Z2480291
The Luer-Lock syringe is free of latex and does not use rubber or silicone lubricant. This product is ideal for IVF, embryo transfer, AI, chromatography and other several applications. The syringe has a volume capacity of 10 mL. It is sold in a pack of 100.