Learn More
Medchemexpress LLC Levacetylleucine | 1188-21-2 | 98.0% | C8H15NO3 | 25 G
SDP
Catalog No. 5000358675
Encompass_Preferred
Levacetylleucine is an orally bioavailable and brain-penetrant compound, derived from the amino acid Leucine. It is the active form of N-acetyl-leucine (NAL). This compound has been shown to attenuate neuronal death and neuroinflammation in mice, and may also improve lysosomal and metabolic dysfunction.
  • Orally bioavailable and brain-penetrant
  • Acetylated derivative of amino acid leucine
  • Active form of N-acetyl-leucine (NAL)
  • Attenuates neuronal death and neuroinflammation
  • Ameliorates lysosomal and metabolic dysfunction
  • Improves postural symptoms after unilateral chemical labyrinthectomy (UL) in rats
  • Promising for research in Niemann-Pick disease type C, traumatic brain injury, and neurodegeneration prevention
Catalog No. 50-003-58675 Supplier Medchemexpress LLC Supplier No. 25GHY59291
Levacetylleucine is an orally bioavailable and brain-penetrant compound, derived from the amino acid Leucine. It is the active form of N-acetyl-leucine (NAL). This compound has been shown to attenuate neuronal death and neuroinflammation in mice, and may also improve lysosomal and metabolic dysfunction.
  • Orally bioavailable and brain-penetrant
  • Acetylated derivative of amino acid leucine
  • Active form of N-acetyl-leucine (NAL)
  • Attenuates neuronal death and neuroinflammation
  • Ameliorates lysosomal and metabolic dysfunction
  • Improves postural symptoms after unilateral chemical labyrinthectomy (UL) in rats
  • Promising for research in Niemann-Pick disease type C, traumatic brain injury, and neurodegeneration prevention