Learn More
Selleck Chemical LLC Levalbuterol tartrate S3153-10mg
SDP
Catalog No. 503109220
Encompass_Preferred
Levalbuterol tartrate(Levosalbutamol tartrate) is a relatively selective beta2-adrenergic receptor agonist used for the treatment of asthma
Catalog No. 50-310-9220 Supplier Selleck Chemical LLC Supplier No. S315310MG
Levalbuterol tartrate(Levosalbutamol tartrate) is a relatively selective beta2-adrenergic receptor agonist used for the treatment of asthma