Learn More
Medchemexpress LLC Levamisole | 14769-73-4 | 99.97% | C11H12N2S | 5 G
SDP
Catalog No. 5000432118
Encompass_Preferred
Levamisole is an analytical standard and anthelmintic agent known for its immunomodulatory properties. It functions as a positive allosteric modulator for the α3β2 and α3β4 nicotinic acetylcholine receptor (nAChR) subtypes. This orally active compound is primarily intended for research and analytical applications.
  • Analytical reference standard
  • Anthelmintic and immunomodulatory agent
  • Positive allosteric modulator for nAChR subtypes
  • Orally active compound
  • Used in qualitative and quantitative research
  • Applied in HPLC, GC, and MS experiments
Catalog No. 50-004-32118 Supplier Medchemexpress LLC Supplier No. HYA01065G
Levamisole is an analytical standard and anthelmintic agent known for its immunomodulatory properties. It functions as a positive allosteric modulator for the α3β2 and α3β4 nicotinic acetylcholine receptor (nAChR) subtypes. This orally active compound is primarily intended for research and analytical applications.
  • Analytical reference standard
  • Anthelmintic and immunomodulatory agent
  • Positive allosteric modulator for nAChR subtypes
  • Orally active compound
  • Used in qualitative and quantitative research
  • Applied in HPLC, GC, and MS experiments