Learn More
Medchemexpress LLC Levamisole hydrochloride | 16595-80-5 | 99.9% | 240.76 | 250 MG
SDP
Catalog No. 5000351130
Encompass_Preferred
Levamisole hydrochloride is an anthelmintic and immunomodulator, classified as a synthetic imidazothiazole derivative. It exhibits antiviral effects against HSV.
  • Anthelmintic
  • Immunomodulator
  • Has antiviral effects against HSV
  • Inhibits most isoforms of alkaline phosphatase (except intestinal and placental)
  • Reduces background alkaline phosphatase activity in biomedical assays
  • Immobilizes the nematode *C. elegans* for imaging
Catalog No. 50-003-51130 Supplier Medchemexpress LLC Supplier No. 250MGHY13666R
Levamisole hydrochloride is an anthelmintic and immunomodulator, classified as a synthetic imidazothiazole derivative. It exhibits antiviral effects against HSV.
  • Anthelmintic
  • Immunomodulator
  • Has antiviral effects against HSV
  • Inhibits most isoforms of alkaline phosphatase (except intestinal and placental)
  • Reduces background alkaline phosphatase activity in biomedical assays
  • Immobilizes the nematode *C. elegans* for imaging