Learn More
Abcam Levamisole hydrochloride, Alkaline phosphatase inhibitor, 250MG
Catalog No. 502706791
Encompass_Preferred
MW 240.75 Da, Purity >99%. Alkaline phosphatase inhibitor. Also a selective agonist at nematode nicotinic acetylcholine receptors (nAChRs). Displays anticancer, immunomodulatory and antiparasitic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6791 Supplier Abcam Supplier No. AB141217250MG
MW 240.75 Da, Purity >99%. Alkaline phosphatase inhibitor. Also a selective agonist at nematode nicotinic acetylcholine receptors (nAChRs). Displays anticancer, immunomodulatory and antiparasitic activity.
The product is subject to the following: Abcam Restricted Use Statement