Learn More
Medchemexpress LLC Levamisole hydrochloride (Standard) | 16595-80-5 | 99.9% | 240.75 | 25 MG
SDP
Catalog No. 5000351458
Encompass_Preferred
Levamisole hydrochloride (Standard) is an analytical and reference standard intended for research and analytical applications. It functions as an anthelmintic and immunomodulator, classified as a synthetic imidazothiazole derivative. Additionally, it exhibits antiviral effects against HSV. This product is for research use only and is not sold to patients.
  • Anthelmintic and immunomodulator
  • Exhibits antiviral effects against HSV
  • Used as an analytical standard for research and analytical applications
  • Functions as a reference standard for assays
  • Applied in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-51458 Supplier Medchemexpress LLC Supplier No. 25MGHY13666R
Levamisole hydrochloride (Standard) is an analytical and reference standard intended for research and analytical applications. It functions as an anthelmintic and immunomodulator, classified as a synthetic imidazothiazole derivative. Additionally, it exhibits antiviral effects against HSV. This product is for research use only and is not sold to patients.
  • Anthelmintic and immunomodulator
  • Exhibits antiviral effects against HSV
  • Used as an analytical standard for research and analytical applications
  • Functions as a reference standard for assays
  • Applied in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS