Learn More
Medchemexpress LLC Levamisole hydrochloride (Standard) | 16595-80-5 | 99.9% | 240.75 | 50 MG
SDP
Catalog No. 5000348764
Encompass_Preferred
Levamisole hydrochloride (Standard) is the analytical standard of Levamisole (hydrochloride), intended for research and analytical applications. It is an anthelmintic and immunomodulator with antiviral effects against HSV. This compound is a reference standard used for qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Levamisole (hydrochloride)
  • Anthelmintic and immunomodulator properties
  • Exhibits antiviral effects against HSV
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-003-48764 Supplier Medchemexpress LLC Supplier No. HY13666R50MG
Levamisole hydrochloride (Standard) is the analytical standard of Levamisole (hydrochloride), intended for research and analytical applications. It is an anthelmintic and immunomodulator with antiviral effects against HSV. This compound is a reference standard used for qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Levamisole (hydrochloride)
  • Anthelmintic and immunomodulator properties
  • Exhibits antiviral effects against HSV
  • Suitable for HPLC, GC, and MS research experiments