Learn More
Medchemexpress LLC Levamlodipine-d4 | 1346617-19-3 | 99.7% | 412.90 | 500 UG
SDP
Catalog No. 5000362276
Encompass_Preferred
Levamlodipine-d4 is the deuterium labeled Levamlodipine. Levamlodipine ((S)-Amlodipine) is a potent dihydropyridine calcium channel blocker that possesses vasodilation properties and is used in the treatment of hypertension and angina.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into the drug molecule, primarily as tracers for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-62276 Supplier Medchemexpress LLC Supplier No. HY14744S500UG
Add to Cart
Add to Cart
Levamlodipine-d4 is the deuterium labeled Levamlodipine. Levamlodipine ((S)-Amlodipine) is a potent dihydropyridine calcium channel blocker that possesses vasodilation properties and is used in the treatment of hypertension and angina.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into the drug molecule, primarily as tracers for quantitation during drug development.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.