Learn More
Abcam Levcromakalim, KATP channel activator, 10MG
Catalog No. 502707562
Encompass_Preferred
MW 286.33 g/mol, Purity >99%. Powerful smooth muscle relaxant. Acts by direct activation of ATP-sensitive K+ channels. Active enantiomer of cromakalim (ab120486). EC₅₀ = 1.6 μM as measured in rat intracardiac ganglia neurones.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7562 Supplier Abcam Supplier No. AB12048510MG
MW 286.33 g/mol, Purity >99%. Powerful smooth muscle relaxant. Acts by direct activation of ATP-sensitive K+ channels. Active enantiomer of cromakalim (ab120486). EC₅₀ = 1.6 μM as measured in rat intracardiac ganglia neurones.
The product is subject to the following: Abcam Restricted Use Statement