Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Level II Polyethylene Isolation Gown
SDP

Catalog No. 19990126
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-990-126 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-990-126 Supplier Keystone Safety Supplier No. ISOTLBLUEA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides a barrier of protection for a variety of industries including healthcare, food, and industrial.

  • Impervious protection against the highest level of fluid
  • Compliant with Title 21 FDA Regulations
  • Produced in an ISO 9001 Certified Factory
  • 100% Latex Free

Specifications

For Use With (Application) Healthcare, Food Manufacturing, Industrial
Width (English) 40 in.
Gender Unisex
Length (English) 47 in.
Size Universal
Product Type General Purpose Gown
No. of Pockets None
Material Polyethylene
Color Blue
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level II, Title 21 FDA Regulations, Conforms to California Proposititon 65
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.