Learn More
Grainger Level Mount Swivel Stud 1/4-20 1in Base
Catalog No. 502750102
Encompass_Preferred
The Level It Male End Leveling Mount Swiveling Pad is a leveling mount for threaded rods and pipes. It features a 1-1/4" diameter swiveling pad that provides 360 degrees of rotation. The mount is constructed of zinc-plated steel and has a weight capacity of 75 lbs. It has a 3/8" - 16 threaded male end to connect to threaded pipe or rod. The swiveling pad allows the mount to adjust to uneven surfaces. This is a high quality leveling mount ideal for leveling threaded rods, pipes, or other applications requiring a swiveling base.
Catalog No. 50-275-0102 Supplier Grainger Supplier No. 14H391
The Level It Male End Leveling Mount Swiveling Pad is a leveling mount for threaded rods and pipes. It features a 1-1/4" diameter swiveling pad that provides 360 degrees of rotation. The mount is constructed of zinc-plated steel and has a weight capacity of 75 lbs. It has a 3/8" - 16 threaded male end to connect to threaded pipe or rod. The swiveling pad allows the mount to adjust to uneven surfaces. This is a high quality leveling mount ideal for leveling threaded rods, pipes, or other applications requiring a swiveling base.