Learn More
Grainger Level Mount Swivel Stud 5/8-11 2-1/2 in.
Catalog No. 502753005
Encompass_Preferred
The S-W Manufacturing Leveling Mount is a heavy duty leveling mount made of cast iron with a weight capacity of 5,800 lbs. It has a 5" rise and drop to allow leveling of equipment. The mount head tilts and rotates 360 degrees. It has a standard 5/8" diameter threaded stem and measures 14" high overall. The leveling mount is designed for leveling heavy machinery and equipment.
Catalog No. 50-275-3005 Supplier Grainger Supplier No. 14H404
The S-W Manufacturing Leveling Mount is a heavy duty leveling mount made of cast iron with a weight capacity of 5,800 lbs. It has a 5" rise and drop to allow leveling of equipment. The mount head tilts and rotates 360 degrees. It has a standard 5/8" diameter threaded stem and measures 14" high overall. The leveling mount is designed for leveling heavy machinery and equipment.