Learn More
Grainger Level Mount Swivel Stud M12 1-7/8in Base
Catalog No. 502753003
Encompass_Preferred
The Level Mount Swivel Stud M12 1-7/8in Base by Level Mount is a swivel stud with a 1-7/8 inch diameter base that can be mounted level. It has an M12 stud size and is made from zinc plated steel for durability. This versatile swivel stud can provide 360 degrees of rotation and has a weight capacity of up to 265 lbs. It can be used for a variety of mounting applications. The stud has a thread reach of 1/2 inch. Overall this is a heavy duty swivel stud that provides flexibility for equipment mounting.
Catalog No. 50-275-3003 Supplier Grainger Supplier No. 14H415
The Level Mount Swivel Stud M12 1-7/8in Base by Level Mount is a swivel stud with a 1-7/8 inch diameter base that can be mounted level. It has an M12 stud size and is made from zinc plated steel for durability. This versatile swivel stud can provide 360 degrees of rotation and has a weight capacity of up to 265 lbs. It can be used for a variety of mounting applications. The stud has a thread reach of 1/2 inch. Overall this is a heavy duty swivel stud that provides flexibility for equipment mounting.