Learn More
Grainger Leveling Legs Metal Gray 4 in H PK2
Catalog No. 502764185
Encompass_Preferred
The Grainger Approved Leveling Legs are made of metal with a gray powder coat finish. They feature a 5/16"-18 threaded stud, providing a load capacity of 250 lbs per leg. The overall height ranges from 2-1/4" to 3-3/4" and they have a 5/8" hex nut for adjusting level. These leveling legs allow equipment to be leveled on uneven surfaces and are ideal for industrial machinery, shelving, workbenches, and other applications needing sturdy, adjustable support. With their sturdy steel construction and wide height adjustment range, these Grainger leveling legs are a versatile option for leveling equipment.
Catalog No. 50-276-4185 Supplier Grainger Supplier No. 36PT89
The Grainger Approved Leveling Legs are made of metal with a gray powder coat finish. They feature a 5/16"-18 threaded stud, providing a load capacity of 250 lbs per leg. The overall height ranges from 2-1/4" to 3-3/4" and they have a 5/8" hex nut for adjusting level. These leveling legs allow equipment to be leveled on uneven surfaces and are ideal for industrial machinery, shelving, workbenches, and other applications needing sturdy, adjustable support. With their sturdy steel construction and wide height adjustment range, these Grainger leveling legs are a versatile option for leveling equipment.