Learn More
Grainger Leveling Mount Boltless M10 1-1/4in Base
Catalog No. 502753006
Encompass_Preferred
The Leveling Mount Boltless M10 1.1/4in Base is a leveling mount with a 1.1/4 inch diameter base that uses an M10 thread. It has a maximum height of 2 inches and is made from zinc plated steel for durability. The leveling mount uses a boltless design for easy height adjustment and can support up to 110 lbs static load. It features a wide base for stability and a hardened steel thread for smooth height adjustment. The leveling mount is ideal for leveling machines, conveyors, signs, gates, and more while providing vibration dampening. It requires no drilling or bolting for installation.
Catalog No. 50-275-3006 Supplier Grainger Supplier No. 14H427
The Leveling Mount Boltless M10 1.1/4in Base is a leveling mount with a 1.1/4 inch diameter base that uses an M10 thread. It has a maximum height of 2 inches and is made from zinc plated steel for durability. The leveling mount uses a boltless design for easy height adjustment and can support up to 110 lbs static load. It features a wide base for stability and a hardened steel thread for smooth height adjustment. The leveling mount is ideal for leveling machines, conveyors, signs, gates, and more while providing vibration dampening. It requires no drilling or bolting for installation.