Learn More
Grainger Leveling Mount Boltless M10 1-1/4in Base
Catalog No. 502750067
Encompass_Preferred
The Grainger Approved Leveling Mount is a heavy duty leveling mount made of steel with a zinc plated finish. It has a weight capacity of up to 5,000 lbs and includes a 5/8" x 4" grade 8 hex head cap screw and lock nut. The leveling mount allows for adjustments of up to 1-1/2" and can handle dynamic, eccentric and vibration loads. This versatile leveling mount provides a stable base for equipment and machinery in industrial environments.
Catalog No. 50-275-0067 Supplier Grainger Supplier No. 14H421
The Grainger Approved Leveling Mount is a heavy duty leveling mount made of steel with a zinc plated finish. It has a weight capacity of up to 5,000 lbs and includes a 5/8" x 4" grade 8 hex head cap screw and lock nut. The leveling mount allows for adjustments of up to 1-1/2" and can handle dynamic, eccentric and vibration loads. This versatile leveling mount provides a stable base for equipment and machinery in industrial environments.