Learn More
Grainger Leveling Mount Boltless M12 1-7/8in Base
Catalog No. 502752071
Encompass_Preferred
The Grainger Approved Leveling Mount is a heavy duty leveling mount made of steel with a zinc plated finish. It has a weight capacity of up to 5,000 lbs and features a 5" x 5" square base plate with four 0.625"-11 threaded holes. The mount height is adjustable from 4.5" to 7" using an Acme thread screw with a 1.5" diameter Acme nut. It can be used for leveling heavy machinery, conveyors, pumps, motors and other equipment. The leveling mount provides vibration dampening and easy equipment adjustment for precision alignment.
Catalog No. 50-275-2071 Supplier Grainger Supplier No. 14H422
The Grainger Approved Leveling Mount is a heavy duty leveling mount made of steel with a zinc plated finish. It has a weight capacity of up to 5,000 lbs and features a 5" x 5" square base plate with four 0.625"-11 threaded holes. The mount height is adjustable from 4.5" to 7" using an Acme thread screw with a 1.5" diameter Acme nut. It can be used for leveling heavy machinery, conveyors, pumps, motors and other equipment. The leveling mount provides vibration dampening and easy equipment adjustment for precision alignment.