Learn More
Grainger Leveling Mount Boltless M16 2-1/2in Base
Catalog No. 502752308
Encompass_Preferred
The Leveling Mount Boltless M16 2-1/2in Base is a leveling mount with a 2-1/2 inch diameter base that allows for 360 degree rotation. It has an M16 threaded stud and is made from steel with a zinc plating. The leveling mount can support up to 265 lbs and allows equipment to be leveled. It can compensate for uneven mounting surfaces. This is an industrial grade leveling mount good for machinery, conveyors, and other equipment that needs leveling.
Catalog No. 50-275-2308 Supplier Grainger Supplier No. 14H429
The Leveling Mount Boltless M16 2-1/2in Base is a leveling mount with a 2-1/2 inch diameter base that allows for 360 degree rotation. It has an M16 threaded stud and is made from steel with a zinc plating. The leveling mount can support up to 265 lbs and allows equipment to be leveled. It can compensate for uneven mounting surfaces. This is an industrial grade leveling mount good for machinery, conveyors, and other equipment that needs leveling.