Learn More
Grainger Leveling Mount Boltless M24 4 in Base
Catalog No. 502752096
Encompass_Preferred
The Leveling Mount Boltless M24 4 in Base from Grainger is a leveling mount with a 4 inch diameter base that allows equipment to be leveled on uneven surfaces. It has a maximum load capacity of 5,000 lbs and is made from cast iron for durability. The leveling mount uses a boltless design with a threaded center rod that adjusts height by rotating the cross bar. It can adjust up to 2 inches above or below level. This heavy duty leveling mount provides sturdy support for large equipment that needs to be leveled.
Catalog No. 50-275-2096 Supplier Grainger Supplier No. 14H431
The Leveling Mount Boltless M24 4 in Base from Grainger is a leveling mount with a 4 inch diameter base that allows equipment to be leveled on uneven surfaces. It has a maximum load capacity of 5,000 lbs and is made from cast iron for durability. The leveling mount uses a boltless design with a threaded center rod that adjusts height by rotating the cross bar. It can adjust up to 2 inches above or below level. This heavy duty leveling mount provides sturdy support for large equipment that needs to be leveled.