Learn More
Grainger LEVEL-IT Leveling Mount, Swivel Boltless, M8 Thread, 25 mm Base
Catalog No. 502753145
Encompass_Preferred
This leveling mount is designed for industrial applications requiring precise height adjustment and stable support. This component features a tapped swivel boltless design with an M8 thread size and a 25 mm (1 in.) diameter round base, providing consistent performance under load.
  • Swivel boltless design allows for a 15° swivel angle
  • Constructed from 303 stainless steel for durability
  • Supports a maximum load capacity of 450 kg
  • Includes a jam nut for secure installation
  • Metric M8 threading
  • Overall height of 17 mm (0.67 in.)
Catalog No. 50-275-3145 Supplier Grainger Supplier No. 14H426
This leveling mount is designed for industrial applications requiring precise height adjustment and stable support. This component features a tapped swivel boltless design with an M8 thread size and a 25 mm (1 in.) diameter round base, providing consistent performance under load.
  • Swivel boltless design allows for a 15° swivel angle
  • Constructed from 303 stainless steel for durability
  • Supports a maximum load capacity of 450 kg
  • Includes a jam nut for secure installation
  • Metric M8 threading
  • Overall height of 17 mm (0.67 in.)