Learn More
Grainger Leveling Mount Swivel Stud M6 3/4in Base
Catalog No. 502753001
Encompass_Preferred
The Leveling Mount Swivel Stud M6 with 3/4in Base from Grainger is a leveling mount with a swivel stud that allows for 360 degree rotation. It has an M6 threaded stud and measures 3/4 inches tall with a 3/4 inch diameter base. The leveling mount is made from zinc-plated steel and can support up to 110 lbs static loads. It features serrated edges on the base for anti-skid and is designed for leveling machinery, conveyors, packaging equipment, and other industrial applications requiring adjustable mounting. This versatile swivel stud mount provides stable, adjustable equipment leveling.
Catalog No. 50-275-3001 Supplier Grainger Supplier No. 14H412
The Leveling Mount Swivel Stud M6 with 3/4in Base from Grainger is a leveling mount with a swivel stud that allows for 360 degree rotation. It has an M6 threaded stud and measures 3/4 inches tall with a 3/4 inch diameter base. The leveling mount is made from zinc-plated steel and can support up to 110 lbs static loads. It features serrated edges on the base for anti-skid and is designed for leveling machinery, conveyors, packaging equipment, and other industrial applications requiring adjustable mounting. This versatile swivel stud mount provides stable, adjustable equipment leveling.