Learn More
Medchemexpress LLC HY-105545A 50mg Medchemexpress, Levetimide CAS:21888-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105545A50MG
Encompass_Preferred
Medchemexpress, HY-105545A 50mg Levetimide CAS:21888-99-3 Levetimide is a potent and stereoselective inhibitor of [3H](+)pentazocine binding, with a Ki of 2.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8275