Learn More
Cayman Chemical Levetiracetam, Purity: ≥98%, CAS: 102767-28-2, 100 mg
SDP
Catalog No. 502894141
Encompass_Preferred
An antiepileptic agent; binds to the synaptic vesicle protein SV2A (IC50 = 1.99 μM); suppresses seizures in fully kindled rats
Catalog No. 50-289-4141 Supplier Cayman Chemical Supplier No. 9001820100 MG
An antiepileptic agent; binds to the synaptic vesicle protein SV2A (IC50 = 1.99 μM); suppresses seizures in fully kindled rats